Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phenylphenol, 99+%, ACROS Organics™

Click to view available options
Quantity:
5g
500g
2.5kg
Packaging:
Glass bottle
Plastic bottle
90-43-7
C12H10O
170.21
MFCD00002208
LLEMOWNGBBNAJR-UHFFFAOYSA-N
2-hydroxybiphenyl,o-phenylphenol,biphenyl-2-ol,2-biphenylol,o-hydroxybiphenyl,2-hydroxydiphenyl,o-hydroxydiphenyl,phenylphenol,biphenylol,1,1'-biphenyl-2-ol
7017
CHEBI:17043
2-phenylphenol
C1=CC=C(C=C1)C2=CC=CC=C2O
90-43-7
C12H10O
170.21
MFCD00002208
LLEMOWNGBBNAJR-UHFFFAOYSA-N
2-hydroxybiphenyl,o-phenylphenol,biphenyl-2-ol,2-biphenylol,o-hydroxybiphenyl,2-hydroxydiphenyl,o-hydroxydiphenyl,phenylphenol,biphenylol,1,1'-biphenyl-2-ol
7017
CHEBI:17043
2-phenylphenol
C1=CC=C(C=C1)C2=CC=CC=C2O
Viscosity 2.4 mPa.s (100°C)
Specific Gravity 1.21
Packaging Plastic bottle
Quantity 500g
Infrared Spectrum Authentic
Melting Point >56.0°C
Boiling Point 282.0°C
Color White
Flash Point 123°C
Density 1.2100g/mL
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Linear Formula C6H5C6H4OH
Beilstein 06,II,623
Merck Index 15,7415
Solubility Information Solubility in water: 0.7g/L(20°C). Other solubilities: readily soluble in naoh solution,ethanol and acet,800g/L isopropanol,soluble in most common organic solvents
Formula Weight 170.21
Physical Form Crystalline Flakes
Percent Purity ≥99%
Chemical Name or Material 2-Phenylphenol, 99+%
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Very toxic to aquatic life.

GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 201-993-5

RTECSNumber : DV5775000

TSCA : TSCA