Learn More
2-Phenylpropionaldehyde, 97%, Thermo Scientific Chemicals
Description
2-phenylpropionaldehyde to give acetophenone using 1 atm molecular oxygen as oxidant was catalyzed by Ce(IV), V(IV) and Bronsted acids. It is used to produce 3-hydroxy-2-methyl-2-phenyl-propionaldehyde. This reaction will need reagent water and K2CO3.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-phenylpropionaldehyde to give acetophenone using 1 atm molecular oxygen as oxidant was catalyzed by Ce(IV), V(IV) and Bronsted acids. It is used to produce 3-hydroxy-2-methyl-2-phenyl-propionaldehyde. This reaction will need reagent water and K2CO3.
Solubility
Soluble in most fixed oils, propylene glycol. Insoluble in glycerin.
Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Density | 1.01 |
Boiling Point | 92°C to 94°C (12 mmHg) |
Flash Point | 79°C (174°F) |
Refractive Index | 1.517 |
Quantity | 100 g |
Beilstein | 4291321 |
Sensitivity | Air Sensitive |
Solubility Information | Soluble in most fixed oils,propylene glycol. Insoluble in glycerin. |
Formula Weight | 134.18 |
Percent Purity | 97% |
Show More |
Safety and Handling
Hazard Category | H227-H315-H317-H319-H361f |
Hazard Statement | GHS H Statement H315-H319-H335-H227 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. Combustible liquid. |
Precautionary Statement | P201-P202-P210-P261-P264b-P272-P280-P281-P302+P352-P305+P351+P338-P308+P313-P333+P313-P362-P370+P378q-P501c |
EINECSNumber | 202-255-5 |
RTECSNumber | CY1460000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.