Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 2-Phenylpropionaldehyde, 97%
Click to view available options
Quantity:
100g
500g
Description
Applications
2-phenylpropionaldehyde to give acetophenone using 1 atm molecular oxygen as oxidant was catalyzed by Ce(IV), V(IV) and Bronsted acids. It is used to produce 3-hydroxy-2-methyl-2-phenyl-propionaldehyde. This reaction will need reagent water and K2CO3.
Solubility
Soluble in most fixed oils, propylene glycol. Insoluble in glycerin.
Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
2-phenylpropionaldehyde to give acetophenone using 1 atm molecular oxygen as oxidant was catalyzed by Ce(IV), V(IV) and Bronsted acids. It is used to produce 3-hydroxy-2-methyl-2-phenyl-propionaldehyde. This reaction will need reagent water and K2CO3.
Solubility
Soluble in most fixed oils, propylene glycol. Insoluble in glycerin.
Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Refractive Index | 1.517 |
Sensitivity | Air Sensitive |
Density | 1.01 |
Flash Point | 79°C (174°F) |
Quantity | 500g |
Boiling Point | 92°C to 94°C (12mmHg) |
Beilstein | 4291321 |
Solubility Information | Soluble in most fixed oils,propylene glycol. Insoluble in glycerin. |
Formula Weight | 134.18 |
Percent Purity | 97% |
Show More |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P201-P202-P210-P261-P264b-P272-P280-P281-P302+P352-P305+P351+P338-P308+P313-P333+P313-P362-P370+P378q-P501c
H227-H315-H317-H319-H361f
EINECSNumber : 202-255-5
RTECSNumber : CY1460000
TSCA : Yes
Recommended Storage : Ambient temperatures