Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phenylpyridine, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
1008-89-5
C11H9N
155.2
MFCD00006280
VQGHOUODWALEFC-UHFFFAOYSA-N
pyridine, 2-phenyl,o-phenylpyridine,pyridine, phenyl,2-phenyl-pyridine,phenyl pyridine,2-azabiphenyl,2-phenyl pyridine,2-phenylpyridine,pubchem13085,2-phenylpyridine????
13887
2-phenylpyridine
C1=CC=C(C=C1)C2=CC=CC=N2
1008-89-5
C11H9N
155.2
MFCD00006280
VQGHOUODWALEFC-UHFFFAOYSA-N
pyridine, 2-phenyl,o-phenylpyridine,pyridine, phenyl,2-phenyl-pyridine,phenyl pyridine,2-azabiphenyl,2-phenyl pyridine,2-phenylpyridine,pubchem13085,2-phenylpyridine????
13887
2-phenylpyridine
C1=CC=C(C=C1)C2=CC=CC=N2
Boiling Point 268°C to 270°C
Flash Point 110°C
Specific Gravity 1.086
Refractive Index 1.6220 to 1.624
Density 1.0860g/mL
Color Colorless to Yellow
Packaging Glass bottle
Quantity 5g
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula (C5H4N)C6H5
Beilstein 20,424
Solubility Information Solubility in water: slightly soluble. Other solubilities: miscible in ethanol,ethyl ether
Formula Weight 155.2
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material 2-Phenylpyridine
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning