Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Phenylquinoline-4-carboxylic acid, 99%

Click to view available options
Quantity:
25g
100g
132-60-5
C16H11NO2
249.269
MFCD00006750
YTRMTPPVNRALON-UHFFFAOYSA-N
cinchophen,tervalon,2-phenyl-4-quinolinecarboxylic acid,cinchophene,cinconal,phenoquin,quinofen,quinophan,quinophen,atofan
8593
2-phenylquinoline-4-carboxylic acid
C1=CC=C(C=C1)C2=NC3=CC=CC=C3C(=C2)C(=O)O
132-60-5
C16H11NO2
249.269
MFCD00006750
YTRMTPPVNRALON-UHFFFAOYSA-N
cinchophen,tervalon,2-phenyl-4-quinolinecarboxylic acid,cinchophene,cinconal,phenoquin,quinofen,quinophan,quinophen,atofan
8593
2-phenylquinoline-4-carboxylic acid
C1=CC=C(C=C1)C2=NC3=CC=CC=C3C(=C2)C(=O)O
Quantity 100g
Odor Odorless
Melting Point 212°C to 218°C
Sensitivity Light sensitive
Beilstein 192803
Merck Index 14,2288
Solubility Information Insoluble in water
Formula Weight 249.27
Percent Purity 99%
Chemical Name or Material 2-Phenylquinoline-4-carboxylic acid

GHS H Statement
H301
Toxic if swallowed.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335-H500

EINECSNumber : 205-067-1

RTECSNumber : GD4025000

TSCA : No

Recommended Storage : Ambient temperatures