Alfa Aesar™ 2-Phenylquinoline, 99+%

Manufacturer: Alfa Aesar™ H3192003

Quantity 1g

 View more versions of this product

Catalog No. AAH3192003

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 612-96-4
    Molecular Formula C15H11N
    Formula Weight 205.26
    MDL Number MFCD00011568
    Chemical Name or Material 2-Phenylquinoline

    Specifications

    Formula Weight 205.26
    Melting Point 82° to 84°C
    Molecular Formula C15H11N
    MDL Number MFCD00011568
    CAS 612-96-4
    Boiling Point 363°C
    Quantity 1g
    Chemical Name or Material 2-Phenylquinoline
    Assay ≥99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit