Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-(Phenylsulfonyl)thiophene, 98%

Manufacturer:  Alfa Aesar™ A1076906

Quantity 5g

 View more versions of this product

Catalog No. AAA1076906


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

22407-40-5
224.3
146279
2- (Phenylsulfonyl)thiophene
C10H8O2S2
MFCD00052142
2-(Benzenesulfonyl)thiophene; Phenyl 2-thienyl sulfone

Specifications

5g
C10H8O2S2
MFCD00052142
2-(Benzenesulfonyl)thiophene; Phenyl 2-thienyl sulfone
2- (Phenylsulfonyl)thiophene
22407-40-5
224.3
146279
121° to 125°C
98%
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit