Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Phenylthio)ethanol, 99%, ACROS Organics™

Click to view available options
Quantity:
10mL
Packaging:
Glass bottle
699-12-7
C8H10OS
154.227
MFCD00002905
KWWZHCSQVRVQGF-UHFFFAOYSA-N
2-phenylthio ethanol,phenylthioethanol,ethanol, 2-phenylthio,2-hydroxyethyl phenyl sulfide,2-phenylmercaptoethanol,2-phenylsulfanyl ethanol,2-phenylthioethanol,unii-9wa374o44p,pubchem10784
69685
2-phenylsulfanylethanol
C1=CC=C(C=C1)SCCO
699-12-7
C8H10OS
154.227
MFCD00002905
KWWZHCSQVRVQGF-UHFFFAOYSA-N
2-phenylthio ethanol,phenylthioethanol,ethanol, 2-phenylthio,2-hydroxyethyl phenyl sulfide,2-phenylmercaptoethanol,2-phenylsulfanyl ethanol,2-phenylthioethanol,unii-9wa374o44p,pubchem10784
69685
2-phenylsulfanylethanol
C1=CC=C(C=C1)SCCO
Color Colorless to Yellow
Quantity 10mL
Refractive Index 1.5900 to 1.5920 (20°C, 589nm)
Packaging Glass bottle
Density 1.14g/mL
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula C6H5SCH2CH2OH
Beilstein 06, II, 291
Formula Weight 154.23
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material 2-(Phenylthio)ethanol, 99%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

STENCH