Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Picoline 98%, ACROS Organics™

Click to view available options
Quantity:
25mL
1L
2.5L
Packaging:
Glass bottle
109-06-8
C6H7N
93.13
MFCD00006332
BSKHPKMHTQYZBB-UHFFFAOYSA-N
2-picoline,o-picoline,pyridine, 2-methyl,picoline,alpha-picoline,a-picoline,pyridine, methyl,methylpyridine,o-methylpyridine,picoline, alpha
7975
CHEBI:50415
2-methylpyridine
CC1=CC=CC=N1
109-06-8
C6H7N
93.13
MFCD00006332
BSKHPKMHTQYZBB-UHFFFAOYSA-N
2-picoline,o-picoline,pyridine, 2-methyl,picoline,alpha-picoline,a-picoline,pyridine, methyl,methylpyridine,o-methylpyridine,picoline, alpha
7975
CHEBI:50415
2-methylpyridine
CC1=CC=CC=N1
Water 0.2% max.
Packaging Glass bottle
Specific Gravity 0.94
Refractive Index 1.4990 to 1.5010
Boiling Point 128.0°C to 129.0°C
Quantity 2.5L
Infrared Spectrum Authentic
Color Yellow
Melting Point -70.0°C
Density 0.9400g/mL
Flash Point 28°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Beilstein 20, 234
Merck Index 15, 7512
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol
Formula Weight 93.13
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 2-Picoline, 98%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 203-643-7

RTECSNumber : TJ4900000

TSCA : TSCA