Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Picolylamine 98.0+%, TCI America™

Click to view available options
Quantity:
25g
250g
3731-51-9
C6H8N2
108.144
MFCD00006360
WOXFMYVTSLAQMO-UHFFFAOYSA-N
2-picolylamine,2-aminomethyl pyridine,2-pyridinemethanamine,2-aminomethylpyridine,2-pyridinemethylamine,2-pyridylmethylamine,2-picolinamine,2-pyridinemethaneamine,2-pyridylmethyl amine,pyridine, 2-aminomethyl
19509
CHEBI:81387
pyridin-2-ylmethanamine
C1=CC=NC(=C1)CN
3731-51-9
C6H8N2
108.144
MFCD00006360
WOXFMYVTSLAQMO-UHFFFAOYSA-N
2-picolylamine,2-aminomethyl pyridine,2-pyridinemethanamine,2-aminomethylpyridine,2-pyridinemethylamine,2-pyridylmethylamine,2-picolinamine,2-pyridinemethaneamine,2-pyridylmethyl amine,pyridine, 2-aminomethyl
19509
CHEBI:81387
pyridin-2-ylmethanamine
C1=CC=NC(=C1)CN
Boiling Point 95°C
Quantity 250g
Color Undesignated
UN Number 2735
Formula Weight 108.14
Physical Form Clear Liquid at 20°C
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material 2-Picolylamine

EINECSNumber : (5)-5596

RTECSNumber : US1840000

TSCA : Yes