Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Piperazine-2-carboxylic acid dihydrochloride, 98%, Acros Organics™

Click to view available options
Quantity:
1g
10g
50g
3022-15-9
C5H12Cl2N2O2
203.063
MFCD00044803
WNSDZBQLMGKPQS-UHFFFAOYSA-N
piperazine-2-carboxylic acid dihydrochloride,2-piperazinecarboxylic acid dihydrochloride,piperazine-2-carboxylic acid 2hcl,+/--piperazine-2-carboxylic acid 2hcl,piperazin-2-carboxylic acid dihydrochloride,2-piperazinecarboxylic acid, dihydrochloride,+/--2-piperazinecarboxylic acid dihydrochloride,+/--piperazine-2-carboxylic acid dihydrochloride,przca-2hcl,r-+-piperazine-2-carboxylic acid dihydrochloride
2723757
piperazine-2-carboxylic acid;dihydrochloride
C1CNC(CN1)C(=O)O.Cl.Cl
3022-15-9
C5H12Cl2N2O2
203.063
MFCD00044803
WNSDZBQLMGKPQS-UHFFFAOYSA-N
piperazine-2-carboxylic acid dihydrochloride,2-piperazinecarboxylic acid dihydrochloride,piperazine-2-carboxylic acid 2hcl,+/--piperazine-2-carboxylic acid 2hcl,piperazin-2-carboxylic acid dihydrochloride,2-piperazinecarboxylic acid, dihydrochloride,+/--2-piperazinecarboxylic acid dihydrochloride,+/--piperazine-2-carboxylic acid dihydrochloride,przca-2hcl,r-+-piperazine-2-carboxylic acid dihydrochloride
2723757
piperazine-2-carboxylic acid;dihydrochloride
C1CNC(CN1)C(=O)O.Cl.Cl
Color White to Yellow
Quantity 1g
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 203.07
Physical Form Crystalline Powder, Crystals and/or Chunks
Percent Purity 97.5 to 102.5%
Chemical Name or Material Piperazine-2-carboxylic acid dihydrochloride

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.