Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Piperazinone, 97%, Alfa Aesar™

Click to view available options
Quantity:
1g
5g
5625-67-2
C4H8N2O
100.121
MFCD01318687
IWELDVXSEVIIGI-UHFFFAOYSA-N
2-piperazinone,2-oxopiperazine,piperazine-2-one,piperazinone,2-piperadone,oxopiperazine,piperazone,keto piperazine,3-oxopiperazine,2-ketopiperazine
231360
piperazin-2-one
C1CNC(=O)CN1
5625-67-2
C4H8N2O
100.121
MFCD01318687
IWELDVXSEVIIGI-UHFFFAOYSA-N
2-piperazinone,2-oxopiperazine,piperazine-2-one,piperazinone,2-piperadone,oxopiperazine,piperazone,keto piperazine,3-oxopiperazine,2-ketopiperazine
231360
piperazin-2-one
C1CNC(=O)CN1
Melting Point 129°C to 134°C
Quantity 1g
Formula Weight 100.12
Percent Purity 97%
Chemical Name or Material 2-Piperazinone

GHS H Statement
H315-H319-H317-H335
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning