Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-piperidinobenzamide, 97%, Maybridge™

Click to view available options
Quantity:
1g
10g
25g
3430-40-8
C12H16N2O
204.27
MFCD00052257
VTXYPPVXMJMLCY-UHFFFAOYSA-N
2-piperidinobenzamide, 2-piperidin-1-yl benzamide, benzamide,2-1-piperidinyl, 2-piperidylbenzamide, piperidinyl benzamide, maybridge1_000974, 2-1-piperidinyl benzamide, benzamide, 2-1-piperidinyl
335067
2-(piperidin-1-yl)benzamide
NC(=O)C1=CC=CC=C1N1CCCCC1
3430-40-8
C12H16N2O
204.27
MFCD00052257
VTXYPPVXMJMLCY-UHFFFAOYSA-N
2-piperidinobenzamide, 2-piperidin-1-yl benzamide, benzamide,2-1-piperidinyl, 2-piperidylbenzamide, piperidinyl benzamide, maybridge1_000974, 2-1-piperidinyl benzamide, benzamide, 2-1-piperidinyl
335067
2-(piperidin-1-yl)benzamide
NC(=O)C1=CC=CC=C1N1CCCCC1
Quantity 25g
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 204.27
Percent Purity 97%
Chemical Name or Material 2-Piperidinobenzamide

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.