Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Piperidylmethyl)-1-cyclohexanone hydrochloride, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
6966-09-2
C12H21NO·HCl
231.77
MFCD00068799
MFMOSYZXKAREGX-UHFFFAOYNA-N
pimeclone hydrochloride,2-piperidylmethyl-1-cyclohexanone hydrochloride,karion respiratory stimulant,1-piperidinomethyl-2-cyclohexanone hydrochloride,2-1-piperidinylmethyl cyclohexanone hydrochloride,2-piperidin-1-ylmethyl cyclohexanone hydrochloride,cyclohexanone, 2-piperidinomethyl-, hydrochloride,2-piperidin-1-ylmethyl cyclohexanone,pimeclone hcl,2-n-piperidinomethyl cyclohexanone hydrochloride
197860
6966-09-2
C12H21NO·HCl
231.77
MFCD00068799
MFMOSYZXKAREGX-UHFFFAOYNA-N
pimeclone hydrochloride,2-piperidylmethyl-1-cyclohexanone hydrochloride,karion respiratory stimulant,1-piperidinomethyl-2-cyclohexanone hydrochloride,2-1-piperidinylmethyl cyclohexanone hydrochloride,2-piperidin-1-ylmethyl cyclohexanone hydrochloride,cyclohexanone, 2-piperidinomethyl-, hydrochloride,2-piperidin-1-ylmethyl cyclohexanone,pimeclone hcl,2-n-piperidinomethyl cyclohexanone hydrochloride
197860
Melting Point 166°C to 170°C
Quantity 1g
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Merck Index 13, 7510
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in methanol,ethanol
Formula Weight 231.77
Percent Purity 98%
Chemical Name or Material 2-(Piperidylmethyl)-1-cyclohexanone hydrochloride, 98%
Show More Show Less

RTECSNumber : GW2080000

TSCA : TSCA