Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Propanethiol 98%, ACROS Organics™

Click to view available options
Quantity:
50mL
500mL
2.5L
Packaging:
Glass bottle
75-33-2
C3H8S
76.15
MFCD00004863
KJRCEJOSASVSRA-UHFFFAOYSA-N
2-propanethiol,isopropyl mercaptan,isopropanethiol,isopropylthiol,2-mercaptopropane,2-propylmercaptan,1-methylethanethiol,isopropylmercaptan,2-propylthiol,2-propane thiol
6364
CHEBI:8474
propane-2-thiol
CC(C)S
75-33-2
C3H8S
76.15
MFCD00004863
KJRCEJOSASVSRA-UHFFFAOYSA-N
2-propanethiol,isopropyl mercaptan,isopropanethiol,isopropylthiol,2-mercaptopropane,2-propylmercaptan,1-methylethanethiol,isopropylmercaptan,2-propylthiol,2-propane thiol
6364
CHEBI:8474
propane-2-thiol
CC(C)S
Flash Point -34°C
Boiling Point 57°C to 60°C
Melting Point -131°C
Specific Gravity 0.82
Refractive Index 1.4245 to 1.4265
Infrared Spectrum Authentic
Density 0.8200g/mL
Color Undesignated
Quantity 2.5L
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula (CH3)2CHSH
Beilstein 01, 367
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohols,ethyl ether,hydrocarbons
Formula Weight 76.15
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material 2-Propanethiol, 98%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 200-861-4

RTECSNumber : TZ7302000

TSCA : TSCA