Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Propanol, anhydrous, >99.5%, Alfa Aesar™

Click to view available options
Quantity:
250mL
1L
4L
67-63-0
C3H8O
60.096
MFCD00011674
KFZMGEQAYNKOFK-UHFFFAOYSA-N
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
3776
CHEBI:17824
propan-2-ol
CC(C)O
67-63-0
C3H8O
60.096
MFCD00011674
KFZMGEQAYNKOFK-UHFFFAOYSA-N
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
3776
CHEBI:17824
propan-2-ol
CC(C)O

Solvent For Sol-Gel Precursors

Water <100ppm
Appearance Colorless liquid
Assay Percent Range >99.5%
Beilstein 635639
Solubility Information Miscible with water,ethanol,acetone,chloroform,methanol,diethyl ether,n-octanol and benzene.
Formula Weight 60.1
Physical Form Liquid
Density 0.785 g/cm3 at 20°C
Ignition Point 455°C (851°F)
Odor Alcohol-like
Refractive Index 1.377
Vapor Pressure 48 hPa (36mm Hg) at 20°C
Flash Point 12°C (53°F)
Boiling Point 82°C to 83°C
Color Colorless
Melting Point °C to 90°C
Quantity 250mL
Chemical Name or Material 2-Propanol
Show More Show Less

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P370+P378q-P501c

H225-H319-H335-H336-H373

DOTInformation : DOT Class: 3, Packaging Group: II

EINECSNumber : 200-661-7

RTECSNumber : NT8050000

TSCA : Yes

Recommended Storage : Ambient temperatures