Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Propanol, anhydrous, 99.5+%, packaged under Argon in resealable ChemSeal™ bottles

Manufacturer:  Alfa Aesar™ 41840K2

Quantity 1L

 View more versions of this product

Catalog No. AA41840K2


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

67-63-0
60.1
UN1219
14,5208
(CH3)2CHOH
MFCD00011674
635639
2-Propanol, Anhydrous

Specifications

67-63-0
Liquid
Alcohol-like
60.1
UN1219
14,5208
Colorless
−90°C
1L
≥99.5%
0.785g/mL
(CH3)2CHOH
ChemSeal™ bottles
MFCD00011674
635639
1.3770
82° to 83°C
12° (53°F)
2-Propanol, Anhydrous

Safety and Handling

SDS & Safety Documents

GHS H Statement
H225-H319-H336
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause drowsiness or dizziness.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOPROPANOL

RTECSNumber : NT8050000

TSCA : TSCA

storageNote1 :

  • Packaged under argon

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit