Alfa Aesar™ 2-Propanol, anhydrous, 99.5+%, packaged under Argon in resealable ChemSeal™ bottles

Manufacturer: Alfa Aesar™ 41840AK

Quantity 250mL

 View more versions of this product

Catalog No. AA41840AK

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 67-63-0
    Linear Formula (CH3)2CHOH
    Formula Weight 60.1
    MDL Number MFCD00011674
    UN Number UN1219
    Beilstein 635639
    Merck Index 14,5208
    Chemical Name or Material 2-Propanol, Anhydrous

    Specifications

    Formula Weight 60.1
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Odor Alcohol-like
    Refractive Index 1.3770
    RTECS Number NT8050000
    Physical Form Liquid
    Packaging ChemSeal™ bottles
    Beilstein 635639
    Health Hazard 1 Danger
    Boiling Point 82° to 83°C
    Melting Point −90°C
    Density 0.785g/mL
    UN Number UN1219
    Merck Index 14,5208
    MDL Number MFCD00011674
    Color Colorless
    Flash Point 12° (53°F)
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOPROPANOL
    TSCA TSCA
    Health Hazard 2 GHS H Statement
    H225-H319-H336
    Highly flammable liquid and vapor.
    Causes serious eye irritation.
    May cause drowsiness or dizziness.
    Quantity 250mL
    Storage Note 1
    • Packaged under argon
    Linear Formula (CH3)2CHOH
    CAS 67-63-0
    Chemical Name or Material 2-Propanol, Anhydrous
    Assay ≥99.5%

    GHS H Statement
    H225-H319-H336
    Highly flammable liquid and vapor.
    Causes serious eye irritation.
    May cause drowsiness or dizziness.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    • Packaged under argon

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit