Learn More
2-Propanol, anhydrous, 99.5+%, Thermo Scientific Chemicals
Description
Solvent for Sol-Gel precursors2-Propanol acts as a solvent for pharmaceuticals, perfumes, gums, resins, alkaloids and essential oils. It is used in cosmetic lotions. Isopropyl alcohol is used as a cleaning agent for keyboards, LCD displays and laptops. It is used as a wetting agent in the fountain solution used in lithographic printing and is often used as a solvent for french polishing shellac, which is used in cabinet making. It acts as a hydride source in the popular Meerwein-Ponndorf-Verley reduction. It is used in huge quantities as a preferred solvent in the manufacture of fine chemicals, drugs, drug intermediates and formulations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | °C to 90°C |
Color | Colorless |
Assay Percent Range | >99.5% |
Appearance | Colorless liquid |
Beilstein | 635639 |
Solubility Information | Miscible with water,ethanol,acetone,chloroform,methanol,diethyl ether,n-octanol and benzene. |
Formula Weight | 60.1 |
Density | 0.785 g/cm3 at 20°C |
Boiling Point | 82°C to 83°C |
Quantity | 250 mL |
Show More |
Safety and Handling
Hazard Category | H225-H319-H335-H336-H373 |
Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P370+P378q-P501c |
DOTInformation | DOT Class: 3, Packaging Group: II |
EINECSNumber | 200-661-7 |
RTECSNumber | NT8050000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.