Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Propanol, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500mL
1L
4L
Packaging:
Aluminum Bottle
HDPE Pressure Bottom Poly Bottle
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC Coated Amber Glass Bottle

Chemical Identifiers

67-63-0
C3H8O
60.096
KFZMGEQAYNKOFK-UHFFFAOYSA-N
3776
CHEBI:17824

High quality chemicals for laboratory and specialized industrial use

    For Laboratory,Research,or Manufacturing Use
SMILES CC(C)O
IUPAC Name propan-2-ol
Formula Weight 60.1
Packaging Aluminum Bottle
Specific Gravity 1L = 0.79kg
Flash Point 12.00°C (53.6°F)
Quantity 4L
Chemical Name or Material 2-Propanol
Grade Baker Analyzed™ ACS Reagent, Meets ACS reagent chemical requirements