Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Propanol, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500mL
1L
4L
Packaging:
Aluminum Bottle
HDPE Pressure Bottom Poly Bottle
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC Coated Amber Glass Bottle
67-63-0
C3H8O
60.096
KFZMGEQAYNKOFK-UHFFFAOYSA-N
isopropanol,2-propanol,isopropyl alcohol,2-hydroxypropane,alkolave,avantine,hartosol,dimethylcarbinol,sec-propyl alcohol,petrohol
3776
CHEBI:17824
propan-2-ol
CC(C)O
67-63-0
C3H8O
60.096
KFZMGEQAYNKOFK-UHFFFAOYSA-N
isopropanol,2-propanol,isopropyl alcohol,2-hydroxypropane,alkolave,avantine,hartosol,dimethylcarbinol,sec-propyl alcohol,petrohol
3776
CHEBI:17824
propan-2-ol
CC(C)O

High quality chemicals for laboratory and specialized industrial use

    For Laboratory,Research,or Manufacturing Use
Formula Weight 60.1
Grade Meets ACS reagent chemical requirements
Packaging Narrow Mouth PVC Coated Amber Glass Bottle
Specific Gravity 1L = 0.79kg
Flash Point 12.00°C (53.6°F)
Quantity 4L
Chemical Name or Material 2-Propanol