Alfa Aesar™ 2-Propanol-d8, 99%(Isotopic)

Manufacturer: Alfa Aesar™ 4231403

Quantity 1g

 View more versions of this product

Catalog No. AA4231403

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 22739-76-0
    Formula Weight 68.15
    MDL Number MFCD00044341
    UN Number UN1219
    Chemical Name or Material 2-Propanol-d8, Isotopic

    Specifications

    Formula Weight 68.15
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Density 0.90g/mL
    Health Hazard 2 GHS H Statement
    H225-H319-H336
    Highly flammable liquid and vapor.
    Causes serious eye irritation.
    May cause drowsiness or dizziness.
    Boiling Point 82.4°C
    Quantity 1g
    Flash Point −11.7°C (53°F)
    UN Number UN1219
    Health Hazard 1 Danger
    Melting Point −89.5°C
    Physical Form Liquid
    MDL Number MFCD00044341
    CAS 22739-76-0
    Odor Alcohol-like
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOPROPANOL
    Chemical Name or Material 2-Propanol-d8, Isotopic
    Assay 99%

    GHS H Statement
    H225-H319-H336
    Highly flammable liquid and vapor.
    Causes serious eye irritation.
    May cause drowsiness or dizziness.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit