Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-Propanol, Filtered through a 0.2 micron filter, BAKER ANALYZED™ LC-MS Reagent, J.T.Baker™
Click to view available options
Quantity:
1L
4L
Packaging:
Narrow Mouth Amber Glass Bottle
Description
- For Use in Liquid Chromatography and Mass Spectrometry; For Laboratory, Research, or Manufacturing Use
Specifications
Formula Weight | 60.1 |
Grade | BAKER ANALYZED™ LC-MS Reagent |
Packaging | Narrow Mouth Amber Glass Bottle |
Specific Gravity | 1L = 0.79kg |
Flash Point | 12.00°C |
Quantity | 4L |
Chemical Name or Material | 2-Propanol, Filtered through a 0.2 micron filter |