Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Propanol, Filtered through a 0.2 micron filter, BAKER ANALYZED™ LC-MS Reagent, J.T.Baker™

Click to view available options
Quantity:
1L
4L
Packaging:
Narrow Mouth Amber Glass Bottle
67-63-0
C3H8O
60.096
KFZMGEQAYNKOFK-UHFFFAOYSA-N
3776
CHEBI:17824
propan-2-ol
CC(C)O
67-63-0
C3H8O
60.096
KFZMGEQAYNKOFK-UHFFFAOYSA-N
3776
CHEBI:17824
propan-2-ol
CC(C)O

Function tested and optimized for LC/MS applications

    For Use in Liquid Chromatography and Mass Spectrometry; For Laboratory, Research, or Manufacturing Use
Formula Weight 60.1
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 0.79kg
Flash Point 12.00°C
Quantity 1L
Grade BAKER ANALYZED™ LC-MS Reagent
Chemical Name or Material 2-Propanol, Filtered through a 0.2 micron filter