Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Propanol, Laboratory Reagent, ≥99.5%, Honeywell™

Click to view available options
Quantity:
1L
2.5L
10L
20L
4 x 2.5L
Packaging:
Glass Bottle
Stainless Steel Can
67-63-0
C3H8O
60.096
MFCD00011674
KFZMGEQAYNKOFK-UHFFFAOYSA-N
isopropanol,2-propanol,isopropyl alcohol,2-hydroxypropane,alkolave,avantine,hartosol,dimethylcarbinol,sec-propyl alcohol,petrohol
3776
CHEBI:17824
propan-2-ol
CC(C)O
67-63-0
C3H8O
60.096
MFCD00011674
KFZMGEQAYNKOFK-UHFFFAOYSA-N
isopropanol,2-propanol,isopropyl alcohol,2-hydroxypropane,alkolave,avantine,hartosol,dimethylcarbinol,sec-propyl alcohol,petrohol
3776
CHEBI:17824
propan-2-ol
CC(C)O

Laboratory Reagent,≥99.5%

  • ≥95% for general laboratory use.
Linear Formula (CH3)2CHOH
UN Number UN1219
Beilstein 635639
Formula Weight 60.1g/mol
Percent Purity ≥99.5%
Grade Laboratory Reagent
Density 0.785g/mL at 25°C
Explosion Limit 0.02
Ignition Point 750°F
Packaging Stainless Steel Can
Refractive Index n20/D 1.377(lit.)
Solubility Soluble in water
Vapor Pressure 33mmHg (20°C), 44mmHg (25°C)
Vapor Density 2.1 (vs air)
Flash Point 12°C (53.6°F)
Physical State Liquid
Boiling Point 77°C to 83°C (1013hPa)
Color Colorless
Melting Point -90°C
Quantity 10L
Chemical Name or Material 2-Propanol
Show More Show Less

P210-P261-P305 + P351 + P338

H225-H319-H336

DOTInformation : Class 3 - PG 2 - Isopropanol

EINECSNumber : 200-661-7

RTECSNumber : NT8050000