Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Propanol, Laboratory Reagent, ≥99.5%, Honeywell™

Catalog Number p-7120079
Click to view available options
Quantity:
1 L
2.5 L
20 L
4 x 2.5 L
Packaging:
Glass Bottle
Stainless Steel Can
67-63-0
C3H8O
60.096
MFCD00011674
KFZMGEQAYNKOFK-UHFFFAOYSA-N
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
3776
CHEBI:17824
propan-2-ol
CC(C)O
67-63-0
C3H8O
60.096
MFCD00011674
KFZMGEQAYNKOFK-UHFFFAOYSA-N
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
3776
CHEBI:17824
propan-2-ol
CC(C)O

Laboratory Reagent,≥99.5%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY
Melting Point -90°C
Color Colorless
Density 0.785g/mL at 25°C
Boiling Point 77°C to 83°C (1013 hPa)
Quantity 2.5 L
Explosion Limit 0.02
Flash Point 12°C (53.6°F)
Ignition Point 750°F
Packaging Glass Bottle
Physical Form Liquid
Refractive Index n20/D 1.377(lit.)
Linear Formula (CH3)2CHOH
Vapor Pressure 33mmHg (20°C), 44mmHg (25°C)
UN Number UN1219
Beilstein 635639
Solubility Information Soluble in water
Formula Weight 60.1g/mol
Percent Purity ≥99.5%
Grade Laboratory Reagent
Chemical Name or Material 2-Propanol
Show More Show Less
Hazard Statement P210-P261-P305 + P351 + P338
Precautionary Statement H225-H319-H336
DOTInformation Class 3 - PG 2 - Isopropanol
EINECSNumber 200-661-7
RTECSNumber NT8050000
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit