Iso-Propanol, Optima™LC/MS Grade, Fisher Chemical Available on GSA/VA Contract for Federal Government customers only.

Packaging Glass Bottle
Quantity 4L

 View more versions of this product

Catalog No. A461-4

  Check Availability
Add to Cart

Custom packing/Bulk ordering

You can now order this item in custom quantities.  Learn More

    Chemical Identifiers

    Additional Information Vapor Pressure: 43mmHg at 20°C
    Aluminum (Al) 10 ppb max.
    Assay Percent Range Assay: ≥99.90%
    Calcium (Ca) 10 ppb max.
    CAS 67-63-0
    Certificate of Analysis Actual Lot Analysis reported on label; Certificate of Actual Lot Analysis available on request
    Copper (Cu) 5 ppb max.
    Density 2.1 (Air = 1.0)
    Boiling Point 81°C
    Color Colorless
    Formula Weight 60.1
    Melting Point −89.5°C
    pH 7
    Physical Form Liquid
    Health Hazard 2 WARNING!
    Health Hazard 3 Emergency Overview
    Flammable liquid and vapor. May cause irritation of respiratory tract. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Hygroscopic. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

    NFPA

    Health:1
    Flammability:3
    Instability:0
    Identification Pass Test
    Iron (Fe) 5 ppb max.
    LCMS Gradient Suitability Pass Test
    Lead (Pb) 5 ppb max.
    Linear Formula (CH3)2CHOH
    Magnesium (Mg) 5 ppb max.
    Manganese (Mn) 5 ppb max.
    Merck Index 15, 5254
    MDL Number 11674
    Molecular Formula C3H8O
    Nickel (Ni) 5 ppb max.
    Packaging Glass Bottle
    Physical State Liquid
    Potassium (K) 10 ppb max.
    Residue after Evaporation 1ppm max.
    Silver (Ag) 5 ppb max.
    Sodium (Na) 50 ppb max.
    Titratable Acid or Base 0.0001 mEq Acid or Base max.
    Vapor Density 2.1 at 20 °C / 68 °F
    Vapor Pressure 43mmHg at 20°C
    Viscosity 2.27 mPaS at 20°C
    Water 0.05% max.
    Zinc (Zn) 10 ppb max.
    Chemical Name or Material 2-Propanol
    Assay ≥99.90%
    Grade Optima™ LC/MS, Also meets ACS specifications

    Specifications

    Boiling Point 81°C
    Melting Point −89.5°C
    Formula Weight 60.1
    Vapor Pressure 43mmHg at 20°C
    Vapor Density 2.1 at 20 °C / 68 °F
    Viscosity 2.27 mPaS at 20°C
    pH 7
    MDL Number 11674
    Quantity 4L
    Merck Index 15, 5254
    Physical Form Liquid
    Color Colorless
    Physical State Liquid
    Certificate of Analysis Actual Lot Analysis reported on label; Certificate of Actual Lot Analysis available on request
    Density 2.1 (Air = 1.0)
    Health Hazard 3 Emergency Overview
    Flammable liquid and vapor. May cause irritation of respiratory tract. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Hygroscopic. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

    NFPA

    Health:1
    Flammability:3
    Instability:0
    Identification Pass Test
    LCMS Gradient Suitability Pass Test
    Lead (Pb) 5 ppb max.
    Nickel (Ni) 5 ppb max.
    Potassium (K) 10 ppb max.
    Zinc (Zn) 10 ppb max.
    Molecular Formula C3H8O
    Linear Formula (CH3)2CHOH
    Absorbance Report AU at 210nm
    CAS 67-63-0
    Synonym IPA
    Iron (Fe) 5 ppb max.
    Additional Information Vapor Pressure: 43mmHg at 20°C
    Magnesium (Mg) 5 ppb max.
    Manganese (Mn) 5 ppb max.
    Packaging Glass Bottle
    Residue after Evaporation 1ppm max.
    Silver (Ag) 5 ppb max.
    Sodium (Na) 50 ppb max.
    Titratable Acid or Base 0.0001 mEq Acid or Base max.
    Water 0.05% max.
    Aluminum (Al) 10 ppb max.
    Assay Percent Range Assay: ≥99.90%
    Calcium (Ca) 10 ppb max.
    Health Hazard 2 WARNING!
    Copper (Cu) 5 ppb max.
    Chemical Name or Material 2-Propanol
    Assay ≥99.90%
    Grade Optima™ LC/MS, Also meets ACS specifications

    WARNING!

    Emergency Overview
    Flammable liquid and vapor. May cause irritation of respiratory tract. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Hygroscopic. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

    NFPA

    Health:1
    Flammability:3
    Instability:0

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Product Certifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit