2-Propanol, Semiconductor Grade, 99.5% min, Alfa Aesar

Maximum level of impurities: Color (APHA) 10, Evaporation residue 5ppm, Solubility in H2O P.T., H2O (by Karl Fischer titrn) 0.05%, Acidity 0.2meq/g, Alkalinity 0.1meq/g, Cl 0.2ppm, PO4 0.5ppm

Manufacturer: Alfa Aesar 19397M1

Quantity 4 x 1L

 View more versions of this product

Catalog No. AA19397M1

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 67-63-0
    Physical Form Liquid
    Assay Percent Range 99.5% min.
    Molecular Formula (CH3)2CHOH
    Formula Weight 60.1
    MDL Number MFCD00011674
    Chemical Name or Material 2-Propanol
    Grade Semiconductor Grade

    Specifications

    Color Clear colorless
    Physical Form Liquid
    Formula Weight 60.1
    Quantity 4 x 1L
    DOT Information DOT Class: 3, Packaging Group: II
    EINECS Number 200-661-7
    Molecular Formula (CH3)2CHOH
    Concentration or Composition (by Analyte or Components) 2-Propanol
    Ignition Point 455°C (851°F)
    Assay Percent Range 99.5% min.
    Odor Alcohol-like
    MDL Number MFCD00011674
    CAS 67-63-0
    Health Hazard 1 H225-H319-H336: Highly flammable liquid and vapour. Causes serious eye irritation. May cause drowsiness or dizziness.
    Vapor Pressure 48 hPa (36mm Hg at 20°C)
    Appearance Colorless liquid
    Chemical Name or Material 2-Propanol
    Grade Semiconductor Grade

    H225-H319-H336: Highly flammable liquid and vapour. Causes serious eye irritation. May cause drowsiness or dizziness.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit