Learn More
2-Propanol, Semiconductor Grade, 99.5% min, Thermo Scientific Chemicals
Description
2-Propanol is used as a solvent for gums, resins, alkaloids and essential oils, as a denaturant for ethyl alcohol and is used in lotions and cosmetics. It is used in keyboard, LCD and laptop cleaning, as a wetting agent in the fountain solution used in lithographic printing, and often used as a solvent for French polishing shellac used in cabinet making. It acts as hydride source in the Meerwein-Ponndorf-Verley reduction. It plays an important role in DNA extraction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 90°C |
Color | Colorless |
Density | 0.785 g/cm3 at 20°C |
Boiling Point | 82°C to 83°C |
Flash Point | 12°C (53°F) |
Ignition Point | 455°C (851°F) |
Odor | Alcohol-like |
Assay Percent Range | 99.5% min. |
Refractive Index | 1.377 |
Quantity | 4 L |
Show More |
Safety and Handling
Hazard Category | H225-H319-H335-H336-H373 |
Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P370+P378q-P501c |
DOTInformation | DOT Class: 3, Packaging Group: II |
EINECSNumber | 200-661-7 |
RTECSNumber | NT8050000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.