Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Propanol (Spectranalyzed™), Fisher Chemical™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
1L
4L
Packaging:
Glass Bottle
67-63-0
C3H8O
60.096
MFCD00011674
KFZMGEQAYNKOFK-UHFFFAOYSA-N
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
3776
CHEBI:17824
propan-2-ol
CC(C)O
67-63-0
C3H8O
60.096
MFCD00011674
KFZMGEQAYNKOFK-UHFFFAOYSA-N
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
3776
CHEBI:17824
propan-2-ol
CC(C)O

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Absorbance Pass Test
Acetone 0.002% max.
Additional Information Vapor Pressure: 43mmHg at 20°C
Assay Percent Range ≥99.5 %
Formula Weight 60.1
Physical Form Liquid
Percent Purity ≥99.5%
Grade Also meets ACS specifications
Density 0.785g/cm³
Identification Pass Test
Infrared Spectrum Conforms to reference
Packaging Glass Bottle
Propionaldehyde 0.002% max.
Residue after Evaporation 0.001% max.
Solubility in Water Pass Test
Titratable Acid or Base 0.0001mEq/g. max.
Vapor Pressure 43mmHg at 20°C
Water 0.2% max.
Viscosity 2.27 mPa/s at 20°C
Alpha Vector Propanol
Boiling Point 81°C
Color Colorless
Melting Point -89.5°C
pH 7
Quantity 1L
Chemical Name or Material 2-Propanol
Show More Show Less

WARNING!

Emergency Overview
Flammable liquid and vapor. May cause irritation of respiratory tract. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Hygroscopic. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:1
Flammability:3
Instability:0

DOTInformation : DOT Class 3, : Flammable Liquid