Alfa Aesar™ 2-Propanol, Spectrophotometric Grade, 99.7+%

Manufacturer: Alfa Aesar™ 39194K7

Quantity 4L

 View more versions of this product

Catalog No. AA39194K7

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 67-63-0
    Formula Weight 60.1
    MDL Number MFCD00011674
    UN Number UN1219
    Beilstein 635639
    Merck Index 14,5208
    Chemical Name or Material 2-Propanol
    Grade Spectrophotometric grade

    Specifications

    Formula Weight 60.1
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H225-H319-H336
    Highly flammable liquid and vapor.
    Causes serious eye irritation.
    May cause drowsiness or dizziness.
    Density 0.785g/mL
    RTECS Number NT8050000
    Odor Alcohol-like
    Boiling Point 82° to 83°C
    Physical Form Liquid
    Melting Point −90°C
    MDL Number MFCD00011674
    CAS 67-63-0
    UN Number UN1219
    Merck Index 14,5208
    Health Hazard 1 Danger
    TSCA TSCA
    Refractive Index 1.3770
    Quantity 4L
    Beilstein 635639
    Flash Point 12° (53°F)
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOPROPANOL
    Chemical Name or Material 2-Propanol
    Assay ≥99.7%
    Grade Spectrophotometric grade

    GHS H Statement
    H225-H319-H336
    Highly flammable liquid and vapor.
    Causes serious eye irritation.
    May cause drowsiness or dizziness.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit