Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Propanol (USP/EP/BP/JP), Fisher Chemical™

Catalog Number A5224
Click to view available options
Quantity:
4 L
20 L
Packaging:
Glass Bottle
Steel Pail
67-63-0
C3H8O
60.096
MFCD00011674
KFZMGEQAYNKOFK-UHFFFAOYSA-N
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
3776
CHEBI:17824
propan-2-ol
CC(C)O
67-63-0
C3H8O
60.096
MFCD00011674
KFZMGEQAYNKOFK-UHFFFAOYSA-N
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
3776
CHEBI:17824
propan-2-ol
CC(C)O

2-Propanol (USP/EP/BP/JP)

TRUSTED_SUSTAINABILITY
pH 7
Melting Point -89.5°C
Absorbance Pass Test (BP), Pass Test (BP/EP), Pass Test (EP)
Color Colorless
Acidity Pass Test (JP), Pass Test (JP/USP), Pass Test (USP)
Appearance Clear and complete (EP)
Density 0.785g/cm³
Boiling Point 81°C
Quantity 4 L
Identification Pass Test (BP), Pass Test (BP/EP/JP/USP), Pass Test (EP), Pass Test (JP), Pass Test (USP)
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.376 to 1.378 (at 20°C) (USP)
Residue after Evaporation 0.005% max. (JP)
Specific Gravity 0.783 to 0.787 (USP), 0.785 to 0.788 (JP), 0.785 to 0.788 (JP), 0.783 to 0.787 (USP)
Linear Formula (CH3)2CHOH
Vapor Pressure 43mmHg at 20°C
Viscosity 2.27 mPa/s at 20°C
Water 0.5% Max. (BP/EP), 0.75% Max. (JP), 0.5% max. (EP), 0.75% max. (JP)
Formula Weight 60.1
Percent Purity ≥99.0% (USP)
Grade BP/EP/FCC/JP/NF/USP
Chemical Name or Material 2-Propanol
Show More Show Less
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. May cause irritation of respiratory tract. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Hygroscopic. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

Note: Products are tested to meet the analytical requirements of the noted grade (USP, Ph. EUR or otherwise). Product test results are available on the Certificate of Analysis or upon request for the specific product.
 
Products are for research use or further manufacturing. Products are not for direct administration to humans
DOTInformation DOT Class 3, : Flammable Liquid
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit