Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Propionylpyrrole, 99%

Click to view available options
Quantity:
1g
5g
1073-26-3
C7H9NO
123.155
MFCD01696449
AEJPPSRYXGEVDT-UHFFFAOYSA-N
2-propionylpyrrole,1-1h-pyrrol-2-yl propan-1-one,2-pyrryl ethyl ketone,ethyl 2-pyrrolyl ketone,alpha-propionylpyrrole,1-2-pyrrolyl-1-propanone,alpha-pyrryl ethyl ketone,1-propanone, 1-pyrrol-2-yl,1-1h-pyrrol-2-yl-1-propanone,unii-hgs11xf57f
61260
1-(1H-pyrrol-2-yl)propan-1-one
CCC(=O)C1=CC=CN1
1073-26-3
C7H9NO
123.155
MFCD01696449
AEJPPSRYXGEVDT-UHFFFAOYSA-N
2-propionylpyrrole,1-1h-pyrrol-2-yl propan-1-one,2-pyrryl ethyl ketone,ethyl 2-pyrrolyl ketone,alpha-propionylpyrrole,1-2-pyrrolyl-1-propanone,alpha-pyrryl ethyl ketone,1-propanone, 1-pyrrol-2-yl,1-1h-pyrrol-2-yl-1-propanone,unii-hgs11xf57f
61260
1-(1H-pyrrol-2-yl)propan-1-one
CCC(=O)C1=CC=CN1
Melting Point 48°C to 51°C
Quantity 5g
Formula Weight 123.15
Percent Purity 99%
Chemical Name or Material 2-Propionylpyrrole

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : UC3805000