Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Propylpentanoic acid, 99%, Thermo Scientific Chemicals

Catalog Number p-188125
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
99-66-1
C8H16O2
144.21
MFCD00002672
NIJJYAXOARWZEE-UHFFFAOYSA-N
valproic acid, dipropylacetic acid, depakene, depakine, 2-propylvaleric acid, ergenyl, di-n-propylacetic acid, valproate, mylproin, pentanoic acid, 2-propyl
3121
CHEBI:39867
2-propylpentanoic acid
CCCC(CCC)C(O)=O
99-66-1
C8H16O2
144.21
MFCD00002672
NIJJYAXOARWZEE-UHFFFAOYSA-N
valproic acid, dipropylacetic acid, depakene, depakine, 2-propylvaleric acid, ergenyl, di-n-propylacetic acid, valproate, mylproin, pentanoic acid, 2-propyl
3121
CHEBI:39867
2-propylpentanoic acid
CCCC(CCC)C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9200g/mL
Boiling Point 220.0°C
Flash Point 111°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4240 to 1.4260
Linear Formula (CH3CH2CH2)2CHCO2H
Quantity 25 g
Beilstein 02, 350
Specific Gravity 0.92
Merck Index 15, 10099
Solubility Information Solubility in water: slightly soluble
Formula Weight 144.21
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Propylpentanoic acid, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye damage.
May damage the unborn child.
Suspected of damaging fertility.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
IF IN EYES: Rinse cautiously with water for several minut
EINECSNumber 202-777-3
RTECSNumber YV7875000
TSCA TSCA

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit