Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Pyridinecarboxaldehyde, 99%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
1121-60-4
C6H5NO
107.11
MFCD00006290
CSDSSGBPEUDDEE-UHFFFAOYSA-N
2-pyridinecarboxaldehyde,picolinaldehyde,pyridine-2-carboxaldehyde,2-formylpyridine,pyridine-2-aldehyde,picolinal,2-picolinaldehyde,2-pyridaldehyde,picolinic aldehyde,2-pyridylaldehyde
14273
CHEBI:73012
pyridine-2-carbaldehyde
C1=CC=NC(=C1)C=O
1121-60-4
C6H5NO
107.11
MFCD00006290
CSDSSGBPEUDDEE-UHFFFAOYSA-N
2-pyridinecarboxaldehyde,picolinaldehyde,pyridine-2-carboxaldehyde,2-formylpyridine,pyridine-2-aldehyde,picolinal,2-picolinaldehyde,2-pyridaldehyde,picolinic aldehyde,2-pyridylaldehyde
14273
CHEBI:73012
pyridine-2-carbaldehyde
C1=CC=NC(=C1)C=O
Flash Point 73°C
Specific Gravity 1.121
Refractive Index 1.5343 to 1.5373
Density 1.1210g/mL
Boiling Point 180.0°C to 182.0°C (761.0mmHg)
Packaging Glass bottle
Quantity 500g
Infrared Spectrum Authentic
Color Brown-Yellow to Yellow
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula (C5H4N)COH
Beilstein 21, I, 287
Solubility Information Solubility in water: miscible. Other solubilities: soluble in most common organic solvents
Formula Weight 107.11
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 2-Pyridinecarboxaldehyde
Show More Show Less

GHS P Statement
Fatal if inhaled.
Harmful if swallowed.
May cause an allergic skin reaction.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Toxic to aquatic life with long lasting effects.

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Avoid release to the environment.
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger