Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Pyrimidinecarbonitrile, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
14080-23-0
C5H3N3
105.1
MFCD00160513
IIHQNAXFIODVDU-UHFFFAOYSA-N
2-cyanopyrimidine,2-pyrimidinecarbonitrile,2-cyano-pyrimidine,2-cyano pyrimidine,cyanopyrimidine,provastatin,pyrimidinecarbonitrile,zlchem 283,pubchem7042,pyrimidin-2-carbonitrile
2757979
pyrimidine-2-carbonitrile
C1=CN=C(N=C1)C#N
14080-23-0
C5H3N3
105.1
MFCD00160513
IIHQNAXFIODVDU-UHFFFAOYSA-N
2-cyanopyrimidine,2-pyrimidinecarbonitrile,2-cyano-pyrimidine,2-cyano pyrimidine,cyanopyrimidine,provastatin,pyrimidinecarbonitrile,zlchem 283,pubchem7042,pyrimidin-2-carbonitrile
2757979
pyrimidine-2-carbonitrile
C1=CN=C(N=C1)C#N
Quantity 5g
Melting Point 37.0°C to 41.0°C
Color White to Yellow
Packaging Glass bottle
Infrared Spectrum Authentic
Flash Point 110°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Solubility Information Solubility in water: .. Other solubilities: soluble in methanol
Formula Weight 105.1
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 2-Pyrimidinecarbonitrile
Show More Show Less

GHS H Statement
Toxic if swallowed.
May cause an allergic skin reaction.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or ras

GHS Signal Word: Danger