Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2-Pyrimidylthio)acetic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
Plastic bottle
88768-45-0
C6H6N2O2S
170.19
MFCD00010287
NIEOYUNNKKAQKI-UHFFFAOYSA-N
2-pyrimidylthio acetic acid,pyrimidin-2-ylthio acetic acid,2-pyrimidin-2-ylthio acetic acid,pyrimidin-2-ylsulfanyl acetic acid,2-pyrimidinylthio acetic acid,acetic acid, 2-pyrimidinylthio,2-carboxymethylthio pyrimidine,2-pyrimidin-2-ylsulfanyl acetic acid,2-pyrimidin-2-ylthioacetic acid,acetic acid, 2-2-pyrimidinylthio
581022
2-pyrimidin-2-ylsulfanylacetic acid
C1=CN=C(N=C1)SCC(=O)O
88768-45-0
C6H6N2O2S
170.19
MFCD00010287
NIEOYUNNKKAQKI-UHFFFAOYSA-N
2-pyrimidylthio acetic acid,pyrimidin-2-ylthio acetic acid,2-pyrimidin-2-ylthio acetic acid,pyrimidin-2-ylsulfanyl acetic acid,2-pyrimidinylthio acetic acid,acetic acid, 2-pyrimidinylthio,2-carboxymethylthio pyrimidine,2-pyrimidin-2-ylsulfanyl acetic acid,2-pyrimidin-2-ylthioacetic acid,acetic acid, 2-2-pyrimidinylthio
581022
2-pyrimidin-2-ylsulfanylacetic acid
C1=CN=C(N=C1)SCC(=O)O
Melting Point 200°C to 203°C
Quantity 5g
Packaging Glass bottle
Infrared Spectrum Authentic
Color Beige to Brown
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 170.19
Physical Form Powder
Percent Purity ≥97.5%
Chemical Name or Material (2-Pyrimidylthio)acetic acid, 98%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 289-445-1

TSCA : TSCA