Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-(Pyrrolidin-1-yl)pyrimidine-5-boronic acid pinacol ester, 95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Specifications
Melting Point | 151.0°C to 152.0°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 94% min. (GC) |
Quantity | 250 mg |
Formula Weight | 275.16 |
Percent Purity | 95% |
Chemical Name or Material | 2-(Pyrrolidin-1-yl)pyrimidine-5-boronic acid pinacol ester |
Safety and Handling
Hazard Category | GHS Signal Word: Warning |
Hazard Statement | GHS H Statement Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell. |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction