Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Quinolinecarboxaldehyde, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
5470-96-2
C10H7NO
157.17
WPYJKGWLDJECQD-UHFFFAOYSA-N
2-quinolinecarboxaldehyde,quinoline-2-carboxaldehyde,2-quinolinecarbaldehyde,2-formylquinoline,quinaldaldehyde,quinolinecarboxaldehyde,quinoline aldehyde,quinoline-2-aldehyde,zlchem 599,pubchem2277
79619
quinoline-2-carbaldehyde
C1=CC=C2C(=C1)C=CC(=N2)C=O
5470-96-2
C10H7NO
157.17
WPYJKGWLDJECQD-UHFFFAOYSA-N
2-quinolinecarboxaldehyde,quinoline-2-carboxaldehyde,2-quinolinecarbaldehyde,2-formylquinoline,quinaldaldehyde,quinolinecarboxaldehyde,quinoline aldehyde,quinoline-2-aldehyde,zlchem 599,pubchem2277
79619
quinoline-2-carbaldehyde
C1=CC=C2C(=C1)C=CC(=N2)C=O
Melting Point 66.0°C to 71.0°C
Color Orange to Brown
Quantity 25g
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Formula Weight 157.17
Physical Form Crystals or Crystalline Powder
Percent Purity 97%
Chemical Name or Material 2-Quinolinecarboxaldehyde
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 226-804-3