Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Quinoxalinecarboxylic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
100mg
Packaging:
Glass bottle
879-65-2
C9H6N2O2
174.16
MFCD00012334
UPUZGXILYFKSGE-UHFFFAOYSA-N
2-quinoxalinecarboxylic acid,2-quinoxalinecarboxylicacid,unii-m5oe8sn42m,2-carboxyquinoxaline,m5oe8sn42m,2-qca,pubchem9776,carbadox metabolite,acmc-1bl7g,quinoxaline carboxylic acid
96695
quinoxaline-2-carboxylic acid
C1=CC=C2C(=C1)N=CC(=N2)C(=O)O
879-65-2
C9H6N2O2
174.16
MFCD00012334
UPUZGXILYFKSGE-UHFFFAOYSA-N
2-quinoxalinecarboxylic acid,2-quinoxalinecarboxylicacid,unii-m5oe8sn42m,2-carboxyquinoxaline,m5oe8sn42m,2-qca,pubchem9776,carbadox metabolite,acmc-1bl7g,quinoxaline carboxylic acid
96695
quinoxaline-2-carboxylic acid
C1=CC=C2C(=C1)N=CC(=N2)C(=O)O
Packaging Glass bottle
Quantity 100mg
Infrared Spectrum Authentic
Melting Point 206.0°C
Color Beige to Green-Yellow
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 174.16
Physical Form Powder, Crystals and/or Chunks
Percent Purity 97%
Chemical Name or Material 2-Quinoxalinecarboxylic acid
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning