Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Quinoxaloyl chloride, 95%, ACROS Organics™

Click to view available options
Quantity:
250mg
1g
54745-92-5
C9H5ClN2O
192.6
SOPDQKNXOCUBSR-UHFFFAOYSA-N
2-quinoxalinecarbonyl chloride,2-quinoxalinecarbonylchloride,2-quinoxaloyl chloride,2-quinoxaloylchloride,2-quinoxalinylcarbonyl chloride,2-quinoxaline carbonyl chloride,2-quinoxalinecarbonyl chloride 7ci, 9ci
2734681
quinoxaline-2-carbonyl chloride
C1=CC=C2C(=C1)N=CC(=N2)C(=O)Cl
54745-92-5
C9H5ClN2O
192.6
SOPDQKNXOCUBSR-UHFFFAOYSA-N
2-quinoxalinecarbonyl chloride,2-quinoxalinecarbonylchloride,2-quinoxaloyl chloride,2-quinoxaloylchloride,2-quinoxalinylcarbonyl chloride,2-quinoxaline carbonyl chloride,2-quinoxalinecarbonyl chloride 7ci, 9ci
2734681
quinoxaline-2-carbonyl chloride
C1=CC=C2C(=C1)N=CC(=N2)C(=O)Cl
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min. (GC)
Formula Weight 192.6
Physical Form Liquid
Percent Purity 95%
Infrared Spectrum Authentic
Packaging Glass Bottle
Color Colorless to Yellow
Melting Point 113.0°C to 115.0°C
Quantity 250mg
Chemical Name or Material 2-Quinoxaloyl chloride
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 259-315-9