Alfa Aesar™ 2-sec-Butyl-4,5-dimethyl-3-thiazoline, 98%

Manufacturer: Alfa Aesar™ H2687009

Quantity 10g

 View more versions of this product

Catalog No. AAH2687009

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 65894-82-8
    Molecular Formula C9H17NS
    Formula Weight 171.3
    MDL Number MFCD01680858
    Chemical Name or Material 2-sec-Butyl-4,5-dimethyl-3-thiazoline

    Specifications

    Formula Weight 171.3
    Molecular Formula C9H17NS
    MDL Number MFCD01680858
    CAS 65894-82-8
    Boiling Point 71.00°C (4.00mmHg)
    Quantity 10g
    Flash Point >70°C (158°F)
    Chemical Name or Material 2-sec-Butyl-4,5-dimethyl-3-thiazoline
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit