Alfa Aesar™ 2-tert-Butyl-4-methylphenol, 99%

Manufacturer: Alfa Aesar™ B2354536

Quantity 500g

 View more versions of this product

Catalog No. AAB2354536

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2409-55-4
    Molecular Formula C11H16O
    Formula Weight 164.25
    MDL Number MFCD00002381
    UN Number UN2430
    Beilstein 1817645
    Synonym 2-tert-Butyl-p-cresol
    Chemical Name or Material 2-tert-Butyl-4-methylphenol

    Specifications

    Formula Weight 164.25
    Molecular Formula C11H16O
    MDL Number MFCD00002381
    Boiling Point 237°C
    Quantity 500g
    Flash Point 105°C (221°F)
    TSCA TSCA
    UN Number UN2430
    Synonym 2-tert-Butyl-p-cresol
    Health Hazard 1 Danger
    Melting Point 49° to 54°C
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ALKYLPHENOLS, SOLID, N.O.S.
    RTECS Number GO7000000
    CAS 2409-55-4
    Beilstein 1817645
    Chemical Name or Material 2-tert-Butyl-4-methylphenol
    Assay 99%

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit