Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-tert-Butylaniline, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass bottle
6310-21-0
C10H15N
149.24
MFCD00130023
AEIOZWYBDBVCGW-UHFFFAOYSA-N
2-tert-butyl aniline,2-tert-butylphenylamine,benzenamine, 2-1,1-dimethylethyl,2-t-butylaniline,2-tert-butyl-phenylamine,2-1,1-dimethylethyl benzenamine,2-tert-butyl phenylamine,pubchem20188,aniline, 2-tert-butyl
80574
2-tert-butylaniline
CC(C)(C)C1=CC=CC=C1N
6310-21-0
C10H15N
149.24
MFCD00130023
AEIOZWYBDBVCGW-UHFFFAOYSA-N
2-tert-butyl aniline,2-tert-butylphenylamine,benzenamine, 2-1,1-dimethylethyl,2-t-butylaniline,2-tert-butyl-phenylamine,2-1,1-dimethylethyl benzenamine,2-tert-butyl phenylamine,pubchem20188,aniline, 2-tert-butyl
80574
2-tert-butylaniline
CC(C)(C)C1=CC=CC=C1N
Boiling Point 123°C to 124°C (17.0mmHg)
Specific Gravity 0.957
Refractive Index 1.5440 to 1.546
Flash Point 102°C
Melting Point -60°C
Packaging Glass bottle
Quantity 25g
Infrared Spectrum Authentic
Color Undesignated
Density 0.9570g/mL
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula (CH3)3CC6H4NH2
Formula Weight 149.24
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 2-tert-Butylaniline, 99%
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Warning

EINECSNumber : 228-634-5