Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-tert-Butylanthraquinone, 95%

Click to view available options
Quantity:
25g
100g
84-47-9
C18H16O2
264.324
MFCD00001232
YTPSFXZMJKMUJE-UHFFFAOYSA-N
2-tert-butylanthraquinone,9,10-anthracenedione, 2-1,1-dimethylethyl,2-tert-butyl-9,10-anthraquinone,2-tert-butyl anthracene-9,10-dione,anthraquinone, 2-tert-butyl,beta-tert-butylanthraquinone,2-t-butylanthraquinone,.beta.-tert-butylanthraquinone,acmc-209puu,2-tert-butyl-anthrachinon
66532
2-tert-butylanthracene-9,10-dione
CC(C)(C)C1=CC2=C(C=C1)C(=O)C3=CC=CC=C3C2=O
84-47-9
C18H16O2
264.324
MFCD00001232
YTPSFXZMJKMUJE-UHFFFAOYSA-N
2-tert-butylanthraquinone,9,10-anthracenedione, 2-1,1-dimethylethyl,2-tert-butyl-9,10-anthraquinone,2-tert-butyl anthracene-9,10-dione,anthraquinone, 2-tert-butyl,beta-tert-butylanthraquinone,2-t-butylanthraquinone,.beta.-tert-butylanthraquinone,acmc-209puu,2-tert-butyl-anthrachinon
66532
2-tert-butylanthracene-9,10-dione
CC(C)(C)C1=CC2=C(C=C1)C(=O)C3=CC=CC=C3C2=O
Melting Point 98°C to 100°C
Quantity 25g
Formula Weight 264.32
Percent Purity 95%
Chemical Name or Material 2-tert-Butylanthraquinone

RTECSNumber : CB0487500

TSCA : TSCA