Alfa Aesar™ 2-tert-Butylanthraquinone, 95%

Manufacturer: Alfa Aesar™ B2363322

Quantity 100g

 View more versions of this product

Catalog No. AAB2363322

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 84-47-9
    Molecular Formula C18H16O2
    Formula Weight 264.32
    MDL Number MFCD00001232
    Chemical Name or Material 2-tert-Butylanthraquinone

    Specifications

    Formula Weight 264.32
    TSCA TSCA
    CAS 84-47-9
    Melting Point 98° to 100°C
    Quantity 100g
    RTECS Number CB0487500
    Molecular Formula C18H16O2
    MDL Number MFCD00001232
    Chemical Name or Material 2-tert-Butylanthraquinone
    Assay 95%

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit