Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-tert-Butylimino-2-diethylamino-1,3-dimethyl-perhydro-1,3,2-diazaphosphorine, 98%, ACROS Organics™

Click to view available options
Quantity:
1mL
5mL
Packaging:
Glass bottle
98015-45-3
C13H31N4P
274.39
VSCBATMPTLKTOV-UHFFFAOYSA-N
bemp,bemp phosphazene,2-tert-butylimino-2-diethylamino-1,3-dimethyl-perhydro-1,3,2-diazaphosphorine,unii-df15j146qi,2-tert-butylimino-2-diethylamino-1,3-dimethylperhydro-1,3,2-diazaphosphorine,2-tert-butylimino-n,n-diethyl-1,3-dimethyl-1,3,2??-diazaphosphinan-2-amine,2-tert-butylimino-n,n-diethyl-1,3-dimethyl-1,3,2,2-t-butylimino-2-diethylamino-1,3-dimethyl-perhydro-1,3,2-diazaphosphorine,2-tert-butylimino-2-diethylamino-1,3-dimethyl-1,3,2-diazaphosphorinane
3513851
2-tert-butylimino-N,N-diethyl-1,3-dimethyl-1,3,2$l^{5}-diazaphosphinan-2-amine
CCN(CC)P1(=NC(C)(C)C)N(CCCN1C)C
98015-45-3
C13H31N4P
274.39
VSCBATMPTLKTOV-UHFFFAOYSA-N
bemp,bemp phosphazene,2-tert-butylimino-2-diethylamino-1,3-dimethyl-perhydro-1,3,2-diazaphosphorine,unii-df15j146qi,2-tert-butylimino-2-diethylamino-1,3-dimethylperhydro-1,3,2-diazaphosphorine,2-tert-butylimino-n,n-diethyl-1,3-dimethyl-1,3,2??-diazaphosphinan-2-amine,2-tert-butylimino-n,n-diethyl-1,3-dimethyl-1,3,2,2-t-butylimino-2-diethylamino-1,3-dimethyl-perhydro-1,3,2-diazaphosphorine,2-tert-butylimino-2-diethylamino-1,3-dimethyl-1,3,2-diazaphosphorinane
3513851
2-tert-butylimino-N,N-diethyl-1,3-dimethyl-1,3,2$l^{5}-diazaphosphinan-2-amine
CCN(CC)P1(=NC(C)(C)C)N(CCCN1C)C
Boiling Point 74°C (0.1mmHg)
Flash Point 53°C
Specific Gravity 0.948
Color Colorless
Density 0.9480g/mL
Quantity 5mL
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Formula Weight 274.39
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 2-tert-Butylimino-2-diethylamino-1,3-dimethyl-perhydro-1,3,2-diazaphosphorine
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapour.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off im

GHS Signal Word: Danger