Alfa Aesar™ (2-tert-Butylphenoxy)acetonitrile, 97+%

Manufacturer: Alfa Aesar™ H52395MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH52395MD

  Check Availability
Add to Cart

    Chemical Identifiers

    Molecular Formula C12H15NO
    Formula Weight 189.26
    MDL Number MFCD00184706
    UN Number UN3276
    Chemical Name or Material (2-tert-Butylphenoxy)acetonitrile

    Specifications

    Formula Weight 189.26
    MDL Number MFCD00184706
    UN Number UN3276
    Health Hazard 3 GHS P Statement
    P261-P280-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Quantity 250mg
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
    Molecular Formula C12H15NO
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302-H312-H332
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Chemical Name or Material (2-tert-Butylphenoxy)acetonitrile
    Assay ≥97%

    GHS H Statement
    H302-H312-H332
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.

    GHS P Statement
    P261-P280-P304+P340-P301+P312-P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit