Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

α,α,α,2-Tetrachloro-6-fluorotoluene, 98%, Acros Organics

Click to view available options
Quantity:
25g
84473-83-6
C7H3Cl4F
247.899
MFCD00000789
PNAGDDZKFJHOOK-UHFFFAOYSA-N
2-chloro-6-fluorobenzotrichloride,1-chloro-3-fluoro-2-trichloromethyl benzene,3-chloro-1-fluoro-2-trichloromethyl benzene,alpha,alpha,alpha,2-tetrachloro-6-fluorotoluene,1-chloro-3-fluoro-2-trichloromethyl benzene #,benzene,1-chloro-3-fluoro-2-trichloromethyl,benzene, 1-chloro-3-fluoro-2-trichloromethyl,1-chloranyl-3-fluoranyl-2-trichloromethyl benzene
611201
1-chloro-3-fluoro-2-(trichloromethyl)benzene
C1=CC(=C(C(=C1)Cl)C(Cl)(Cl)Cl)F
84473-83-6
C7H3Cl4F
247.899
MFCD00000789
PNAGDDZKFJHOOK-UHFFFAOYSA-N
2-chloro-6-fluorobenzotrichloride,1-chloro-3-fluoro-2-trichloromethyl benzene,3-chloro-1-fluoro-2-trichloromethyl benzene,alpha,alpha,alpha,2-tetrachloro-6-fluorotoluene,1-chloro-3-fluoro-2-trichloromethyl benzene #,benzene,1-chloro-3-fluoro-2-trichloromethyl,benzene, 1-chloro-3-fluoro-2-trichloromethyl,1-chloranyl-3-fluoranyl-2-trichloromethyl benzene
611201
1-chloro-3-fluoro-2-(trichloromethyl)benzene
C1=CC(=C(C(=C1)Cl)C(Cl)(Cl)Cl)F
Melting Point 38°C to 43°C
Color White
Quantity 25g
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula ClC6H3(F)CCl3
Formula Weight 247.91
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material α,α,α,2-Tetrachloro-6-fluorotoluene, 98%
Show More Show Less