Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Tetrahydrofurfuryloxy)tetrahydropyran 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
710-14-5
C10H18O3
186.251
MFCD00006603
XXNUDHUOMVIIFN-UHFFFAOYSA-N
2-tetrahydrofurfuryloxy tetrahydropyran,2 3-tetrahydrofurfuryloxy tetrahydropyran,2-oxolan-2-ylmethoxy oxane,tetrahydro tetrahydrofuryl methoxy-2h-pyran,2h-pyran, tetrahydro-2-tetrahydrofurfuryl oxy,2h-pyran, tetrahydro-2-tetrahydro-2-furanyl methoxy,acmc-209soh,tetrahydrofurfuroxy tetrahydropyran,2-tetrahydrofurfuroxy tetrahydropyran,2 3-tetrahydrofurfuryloxy tetrahydro-pyran
97985
2-(oxolan-2-ylmethoxy)oxane
C1CCOC(C1)OCC2CCCO2
710-14-5
C10H18O3
186.251
MFCD00006603
XXNUDHUOMVIIFN-UHFFFAOYSA-N
2-tetrahydrofurfuryloxy tetrahydropyran,2 3-tetrahydrofurfuryloxy tetrahydropyran,2-oxolan-2-ylmethoxy oxane,tetrahydro tetrahydrofuryl methoxy-2h-pyran,2h-pyran, tetrahydro-2-tetrahydrofurfuryl oxy,2h-pyran, tetrahydro-2-tetrahydro-2-furanyl methoxy,acmc-209soh,tetrahydrofurfuroxy tetrahydropyran,2-tetrahydrofurfuroxy tetrahydropyran,2 3-tetrahydrofurfuryloxy tetrahydro-pyran
97985
2-(oxolan-2-ylmethoxy)oxane
C1CCOC(C1)OCC2CCCO2
Density 1.03g/mL
Quantity 25g
Color Colorless
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4600 to 1.4620 (20°C, 589nm)
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 186.25
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material 2-(Tetrahydrofurfuryloxy)tetrahydropyran, 98%
Show More Show Less

EINECSNumber : 211-917-2

TSCA : TSCA